Learn More
Bioss p38 MAP Kinase - C-terminal A
SDP
Supplier:  Bioss BSM70584M
Encompass_Preferred
p38 MAP Kinase (C-terminal) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M548Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65670