Learn More
Selleck Chemical LLC p38-MAPK-IN-1 E2372-25MG
SDP
Supplier:  Selleck Chemical LLC E237225MG
Encompass_Preferred
p38- MAPK-IN-1 is an inhibitor of p38- with IC50 value of 2300 nM in EFC displacement assay and 5500 nM in HTRF assay
Catalog No. 50-000-40800