Learn More
Medchemexpress LLC HY-12839 5mg Medchemexpress, p38 MAPK-IN-1 CAS:1006378-90-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128395MG
Encompass_Preferred
Medchemexpress, HY-12839 5mg p38 MAPK-IN-1 CAS:1006378-90-0 p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1166