Learn More
ECM Bioscience p38 MAPK Phospho-Regulation Immunocytochemistry Kit, ECM Biosciences
Supplier:  ECM Bioscience PK7690
Encompass_Preferred
The p38 MAPK phospho-regulation kit can be used for immunocytochemical co-localization of p38 MAPK phosphorylation at Thr-180/Tyr-182 and Tyr-323 relative to the total expression of p38 MAPK. The kit includes rabbit polyclonal and mouse monoclonal antibodies along with Goat-anti-Rabbit conjugated to DyLight 594 and Goat anti-Mouse conjugated to DyLight 488 for dual labeling experiments. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8784