Learn More
Cayman Chemical p38a MAPK-IN-1 50mg
SDP
Supplier:  Cayman Chemical 4083950 MG
Encompass_Preferred
A p38α MAPK inhibitor (IC50 = 5,500 nM in an HTRF assay); inhibits LPS-induced TNF-α production in THP-1 cells (EC50 = 6,200 nM); has been used in the study of p38α MAPK inhibitor binding interactions and kinetics
Catalog No. 50-294-3479