Learn More
Electron Microscopy Sciences P47 Scintillator for Cambridge S600, 12.0 x 1 mm
SDP
Supplier:  Electron Microscopy Sciences 82017
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Cambridge S600.
Size: 12.0 x 1 mm
Catalog No. 50-276-12