Learn More
Medchemexpress LLC P5(PEG24)-VC-PAB-Exatecan | 2928571-43-9 | 99.7% | 2105.28 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1630991MG
Encompass_Preferred
P5(PEG24)-VC-PAB-Exatecan (LP5) is a drug-linker conjugate composed of a potent DNA Topoisomerase I inhibitor Exatecan (HY-13631) and a linker P5(PEG24)-VC-PAB to make antibody agent conjugate (ADC). It can be used for tumor research.
  • Drug-linker conjugate for ADC
  • Contains a potent DNA Topoisomerase I inhibitor (Exatecan)
  • Can be used for the research of tumor
Catalog No. 50-003-91618