Learn More
Selleck Chemical LLC P5091 (P005091) 10mg 882257-11-6
SDP
Supplier:  Selleck Chemical LLC S713210MG
Encompass_Preferred
P5091(P005091) is a selective and potent inhibitor of ubiquitin-specific protease 7 (USP7) with EC50 of 4.2 ?M and the closely related USP47. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4437