Learn More
Enzo Life Sciences p53 Recombinant monoclonal antibody (PAb421) (Mouse IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6530200
Encompass_Preferred
Clone: PAb421. Isotype: IgG2aκ. Immunogen: Synthetic peptide corresponding to aa 371-380 of human p53. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey. Specificity: Recognizes the mammalian mutant and wild-type p53 protein and reacts with an epitope between aa 370-378 (near the C-terminus of the protein). Applications: ELISA, IF, IHC (FS), IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3177