Learn More
Proteintech Group Inc P63 RABBIT POLYAB
SDP
Supplier:  Proteintech Group Inc 121431AP150UL
Encompass_Preferred
Host Species: Rabbit Cross Reactivity: Human,Mouse,Rat Immunogen: Recombinant Protein, C-term-300 amino acid Format:Antigen affinity purification Application: ELISA, WB Recommended Storage: - 20 deg.C or lower. This antibody has been cited in 1 publications, with application WB to species rat. PMID: 25136779; Yun-Guo Wang et al. , All-trans-retinoid acid (ATRA) suppresses chondrogenesis of rat primary hind limb bud mesenchymal cells by downregulating p63 and c
Catalog No. 50-561-166