Learn More
Medchemexpress LLC P70 S6 Kinase beta/SRK (phospho S371) Antibody (YA1890) | 100 UL
SDP
Supplier:  Medchemexpress LLC HYP82145100UL
Encompass_Preferred
This antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target P70 S6 Kinase beta/SRK (phospho S371). It is suitable for use in Western blot (WB) applications and demonstrates reactivity with human, mouse, and rat samples.
  • Rabbit-derived host
  • Non-conjugated IgG monoclonal antibody
  • Targets P70 S6 Kinase beta/SRK (phospho S371)
  • Reactive with human, mouse, and rat
  • Suitable for Western blot applications
Catalog No. 50-004-41521