Learn More
Enzo Life Sciences p70 S6K activity kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS470
Encompass_Preferred
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio. Use/Stability: Store all components at 4°C, except active kinase at -80°C. UniProt ID: P23443 (S6K1), Q9UBS0 (S6K2). Applications: Colorimetric detection Activity assay. Species reactivity: Species independent . Content: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2.
Catalog No. 50-200-7214