Learn More
Medchemexpress LLC PA452 | 457657-34-0 | ≥99.0% | 439.59 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1085221MG
Encompass_Preferred
PA452 is a specific antagonist of the retinoic X receptor (RXR) that inhibits the effect of Retinoic acid (RA) on Th1/Th2 development. In vitro, PA452 inhibits Troglitazone (TZ)-induced CK13 expression and inhibits RXR in normal human urothelial (NHU) cells.
  • Specific antagonist of the retinoic X receptor (RXR)
  • Inhibits the effect of retinoic acid (RA) on Th1/Th2 development
  • Inhibits Troglitazone (TZ)-induced CK13 expression
  • Inhibits RXR in normal human urothelial (NHU) cells
Catalog No. 50-003-46831