Learn More
Cayman Chemical PAC1 Fatty Acids and Derivativ
SDP
Supplier:  Cayman Chemical 1000931725 MG
Encompass_Preferred
A procaspase-3 activator and a potential drug treatment in cancer cell lines with elevated levels of procaspase-3; exhibits an IC50 value of 3 nM for induction of cancer cell death without affecting non-cancerous cells
Catalog No. 50-289-0912