Learn More
Medchemexpress LLC PACAP (1-27), human, ovine, rat | 127317-03-7 | 99.3% | 3147.66 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP0176500UG
Encompass_Preferred
PACAP (1-27), human, ovine, rat (PACAP 1-27) is the N-terminal fragment of PACAP-38. This peptide functions as a potent PACAP receptor agonist, making it suitable for various research applications.
  • Functions as a potent PACAP receptor agonist with specific IC50s for rat PAC1 (3 nM), rat VPAC1 (2 nM), and human VPAC2 (5 nM).
  • Demonstrated inhibitory effects on increased total pulmonary resistance caused by allergens or histamine in guinea pigs.
  • Exhibits high solubility, dissolving at 100 mg/mL in DMSO and ≥ 50 mg/mL in water.
  • Stable for 2 years as a powder when stored at -80°C; up to 6 months in solvent at -80°C in sealed storage away from moisture.
Catalog No. 50-004-26079