Learn More
Echelon Biosciences Research Labs PACAP (1-27), ovine, human / Adenylate Cyclase Activating Polypeptide 27, ovine, human - 1 mg, CAS#: 127317-03-7
SDP
Supplier:  Echelon Biosciences Research Labs 350371MG
Encompass_Preferred
PACAP (1-27), ovine, human / Adenylate Cyclase Activating Polypeptide 27, ovine, human - 1mg , His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8415