Learn More
Medchemexpress LLC PACAP (1-38) free acid | 129405-61-4 | 99.9% | 4535.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0221C5MG
Encompass_Preferred
PACAP (1-38) free acid is a desamido-PACAP (1-38), which is human, ovine, and rat. It activates the human PAC1 receptor and induces cyclic AMP (cAMP) generation in both NS-1 neuroendocrine cells and non-neuroendocrine HEK293 cells. In vitro, it promotes neuroendocrine cell differentiation in NS-1 cells with an EC50 of 10.6 nM and induces cell growth arrest in NS-1 cells at concentrations of 0.1-300 nM.
  • Activates human PAC1 receptor
  • Induces cyclic AMP (cAMP) generation in NS-1 neuroendocrine cells
  • Induces cyclic AMP (cAMP) generation in non-neuroendocrine HEK293 cells
  • Promotes neuroendocrine cell differentiation in NS-1 cells
  • Induces cell growth arrest in NS-1 cells
Catalog No. 50-004-25992