Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC Pacific Biosciences Paramagnetic Beads, SMRT Sequencing Workflow, Research Use Only
Supplier: PACIFIC BIOSCIENCES OF CALIFORNIA INC 100265900
AMPure PB contains specially formulated paramagnetic beads for the SMRT™ sequencing workflow. This liquid suspension, appearing as brown beads in a clear liquid, is intended for research use only.
- Specially formulated paramagnetic beads for SMRT™ sequencing workflow.
- Appearance is brown beads suspended in clear liquid.
- Beads may settle to the bottom of the bottle.
- No odor.
- pH of 8.0 to 8.4.
- Relative density of ˜1.127.
- Fully miscible in water.
- Contains Sodium azide < 0.1%.
- Shipped at ambient temperature (15°C to 30°C) and stored refrigerated (2°C to 8°C) upon receipt.
- Mix reagent well before use to ensure homogenous and consistent color.
- Intended for research use only, not for diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.