Learn More
Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.
 View more versions of this product
Catalog No. 022924
Item Discontinued This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:
  • 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):
  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors
KONTACT (Particulate Activator) (Product No. 01-0402):
  • Higher sensitivity to Factor VIII
  • Average Normal Values: 26 - 33 Seconds
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions