Learn More
Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.
 View more versions of this product
Catalog No. 02-292-9
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:
  • 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):
  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors
KONTACT (Particulate Activator) (Product No. 01-0402):
  • Higher sensitivity to Factor VIII
  • Average Normal Values: 26 - 33 Seconds
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions