Learn More
AdipoGen Paclitaxel,Chemical. CAS: 33069-62-4
SDP
Supplier:  AdipoGen AGCN20045M100
Encompass_Preferred
Paclitaxel, AdipoGen Life Sciences. Chemical. CAS: 33069-62-4. Formula: C47H51NO14. MW: 853.9. Isolated from the bark of the pacific yew tree (Taxus brevifolia). Anticancer compound. Chemotherapeutic used in patients with cancer and advanced forms of Kaposi's sarcoma. Microtubule assembly stabilizer. Reversibly binds to polymerized tubulin. Anti-mitotic. Mitotic spindle assembly, chromosome segregation and cell division inhibitor. Induces cell cycle arrest at the G2/M phase. Apoptosis inducer through aberrant activation of cyclin-dependent kinases (CKDs) and the c-Jun N-terminal kinase/stress activated protein kinase (JNK/SAPK). Immunosuppressor. Immunostimulant. Antiproliferative agent for the prevention of restenosis. TRAIL sensitizer 33069-62-4
Catalog No. 50-117-5920