Learn More
Medchemexpress LLC HY-B0853 250mg Medchemexpress, Paclobutrazol CAS:76738-62-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0853250MG
Encompass_Preferred
Medchemexpress, HY-B0853 250mg Paclobutrazol CAS:76738-62-0 Paclobutrazol is a triazole-containing plant growth retardant that is known to inhibit the biosynthesis of gibberellins. Paclobutrazol also has antifungal activities. Paclobutrazol, transported acropetally in plants, can also suppress the synthesis of abscisic acid and induce chilling tolerance in plants. Paclobutrazol is typically used to support research on the role of gibberellins in plant biology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2667