Learn More
Medchemexpress LLC Paclobutrazol | 76738-62-0 | 97.1% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB08531G
Encompass_Preferred
Paclobutrazol is a triazole-containing plant growth retardant and an analytical standard. It is known for its ability to inhibit the biosynthesis of gibberellins and exhibits antifungal activities. This compound is transported acropetally in plants, where it can suppress the synthesis of abscisic acid, leading to induced chilling tolerance. It is primarily used in research to investigate the role of gibberellins in plant biology.
  • Inhibits the biosynthesis of gibberellins
  • Possesses antifungal properties
  • Can suppress abscisic acid synthesis
  • Induces chilling tolerance in plants
  • Used as an analytical standard
  • Valuable tool for research on plant biology
Catalog No. 50-004-07955