Learn More
Medchemexpress LLC Paclobutrazol | 76738-62-0 | 97.1% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB085310MM/1ML
Encompass_Preferred
Paclobutrazol is a triazole-containing plant growth retardant that is known to inhibit the biosynthesis of gibberellins. It also has antifungal activities, can suppress the synthesis of abscisic acid, and induces chilling tolerance in plants. Transported acropetally, it is typically used to support research on the role of gibberellins in plant biology.
  • Inhibits biosynthesis of gibberellins
  • Possesses antifungal activities
  • Suppresses synthesis of abscisic acid
  • Induces chilling tolerance in plants
  • Used for research on gibberellins in plant biology
  • Functions as a triazole-containing plant growth retardant
  • Transported acropetally in plants
Catalog No. 50-004-07954