Learn More
Medchemexpress LLC Paclobutrazol | 76738-62-0 | 97.38% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0853500MG
Encompass_Preferred
Paclobutrazol is a triazole-containing plant growth retardant that inhibits the biosynthesis of gibberellins and possesses antifungal activities. It is transported acropetally in plants, where it can suppress the synthesis of abscisic acid and induce chilling tolerance. This substance is commonly utilized in research to study the role of gibberellins in plant biology.
  • Increases total soluble sugars, starch, protein, and free amino acids in mature Brassica juncea seeds.
  • Inhibits the growth of various fungi, including Sirococcus clavigignenti-juglandacearum and Ceratocystis fagacearum.
  • Improves the quality of tomato seedlings and enhances resistance to early blight disease.
Catalog No. 50-004-08899