Learn More
Medchemexpress LLC Paclobutrazol (Standard) | 76738-62-0 | 96.55% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0853R50MG
Encompass_Preferred
Paclobutrazol (Standard) is the analytical standard of Paclobutrazol, intended for research and analytical applications. This compound acts as a triazole-containing plant growth retardant by inhibiting gibberellin biosynthesis and also exhibits antifungal activities. It can suppress abscisic acid synthesis and induce chilling tolerance in plants when transported acropetally.
  • Inhibits gibberellin biosynthesis
  • Exhibits antifungal activities
  • Suppresses abscisic acid synthesis
  • Induces chilling tolerance in plants
  • Reference standard for qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-08859