Learn More
Medchemexpress LLC Pacma 31 | 1401089-31-3 | 99.3% | 430.47 g·mol⁻1 | C21H22N2O6S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100433100MG
Encompass_Preferred
PACMA 31 is an irreversible small-molecule inhibitor of protein disulfide isomerase (PDI) used in biochemical and cellular research. It covalently modifies active-site cysteines, has shown tumor-targeting activity in preclinical studies, and contains an alkyne moiety that enables copper-catalyzed azide-alkyne cycloaddition for conjugation and probe development.
  • Irreversible inhibitor of protein disulfide isomerase for biochemical and cellular assays.
  • Forms covalent bond with active-site cysteines for sustained target engagement.
  • Contains an alkyne group suitable for click chemistry conjugation.
  • High purity suitable for research use.
  • Available in multiple pack sizes for flexible experimental needs.
Catalog No. 50-002-14160