Learn More
Medchemexpress LLC Pacma 31 | 1401089-31-3 | MFCD26793891 | 99.3% | 430.47 g/mol | C21H22N2O6S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10043310MM 1ML
Encompass_Preferred
Pacma 31 is an irreversible protein disulfide isomerase (PDI) inhibitor supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It covalently modifies active-site cysteines in PDI and is used in biochemical, cellular, and in vivo studies to probe PDI-dependent redox folding and tumor biology.
  • Irreversible PDI inhibition mechanism for target validation.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High reported purity suitable for biochemical assays.
  • Supplied in a 1 mL format for small-volume experiments.
  • CAS number provided for unambiguous chemical identification.
Catalog No. 50-002-15473