Learn More
Selleck Chemical LLC Pacritinib (SB1518) 5mg 937272-79-2
SDP
Supplier:  Selleck Chemical LLC S80575MG
Encompass_Preferred
Pacritinib (SB1518) is a potent and selective inhibitor of Janus Kinase 2 (JAK2) and Fms-Like Tyrosine Kinase-3 (FLT3) with IC50s of 23 and 22 nM in cell-free assays, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0666942