Learn More
Molecular Dimensions PACT PREMIUM MD1-36
SDP
Supplier:  Molecular Dimensions MD136
Encompass_Preferred
Protein Crystallography. PACT premier screen HT-96 (96 x 1.1mL) is 3 screens in one box: a 24 well pH/PEG screen - with broad range buffers thus maintaining consistent buffer chemistry across the pH range, a 24 well Cation/PEG screen and a 48 well anion/PEG screen.
Catalog No. NC0213660