Learn More
Medchemexpress LLC Pactimibe sulfate | 608510-47-0 | 98.0% | 465.65 | C25H40N2O3.1/2H2O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100401A10MM 1ML
Encompass_Preferred
Pactimibe sulfate is a small-molecule dual ACAT1/2 inhibitor used in biochemical and cellular research to investigate cholesterol esterification and related pathways. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it has reported low-micromolar activity in enzyme and cell-based assays and is provided at high purity for research applications.
  • Ready-to-use 10 mM DMSO solution for biochemical and cellular assays.
  • Dual ACAT1/2 inhibitory activity with reported IC50s in the low micromolar range.
  • High purity (98.0%) appropriate for research use.
  • Suitable for studies of cholesterol esterification and atherosclerosis-related mechanisms.
  • Provided in a small-volume solution convenient for screening and dose-response experiments.
Catalog No. 50-002-14857