Learn More
Beckman Coulter GH 3/8 Adapter Rubber Pad, 15ml, 14 Tube, 349949
Supplier:  Beckman Coulter 349949
Encompass
The Beckman GH 3/8 Adapter Rubber Pad is designed for use with 15mL (14mm diameter) centrifuge tubes. It features a 3/8 inch diameter opening and is made of rubber to provide a secure, non-slip grip. This rubber adapter pad can accommodate a single 15mL tube and is compatible with Beckman Coulter centrifuges that accept 3/8 inch accessories. It provides safe centrifugation by securing 15mL tubes to prevent breakage. This is an OEM replacement part, manufactured for use in Beckman Coulter centrifuges.
Catalog No. NC1021888