Learn More
Med Vet International Antiseptic Prep Pad Prevantics 3.15% / 70% Strength CHG (Chlorhexidine Gluconate) / Isopropyl Alcohol Individual Packet NonSterile, 1ML, B10800, Each
Supplier:  Med Vet International 532824EA
Encompass_Preferred
Pre-injection skin preparation Achieves >2 log10 in 30 seconds and maintains 2 log10 bacterial reduction for at least 24 hours Easy-to-use prep pad design covers a 6.25 square inch treatment area
Catalog No. 50-304-4473