Learn More
Cayman Chemical PAD3 MoncLNal ANTIBDYClo 100ug
SDP
Supplier:  Cayman Chemical 24377100 UG
Encompass_Preferred
Immunogen: Full length recombinant PAD3 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) PAD3; (-) PAD1, PAD2, PAD4 • Applications: ELISA, IF, WB • MW = ~74 kDa
Catalog No. 50-293-3857