Learn More
Medchemexpress LLC Padsevonil | 1294000-61-5 | 99.7% | 432.80 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1090091MG
Encompass_Preferred
Padsevonil (UCB0942) is a potent antiepileptic agent that selectively acts on presynaptic and postsynaptic targets. It binds to synaptic vesicular protein 2 (SV2) with high affinity and is also a positive allosteric modulator and partial agonist of GABAAR, showing high potency against α1 and α5 receptors.
  • Regulates GABAAR activity through allosteric sites on the receptor in CHO cells.
  • Enhances natural GABAAR activity in primary rat neurons.
  • Has protective effects on mouse models of acute epilepsy.
  • Shows a protective effect in a rodent model of chronic epilepsy.
  • Significantly decreased hippocampal paroxysmal discharges dose-dependently in kainate treated male mice.
  • Produced dose-related suppression in the cumulative duration of spontaneous spike and wave discharges in platinum electrodes treated male Wistar rats.
Catalog No. 50-003-46648