Learn More
Medchemexpress LLC HY-N7686 1mg Medchemexpress, Paeoniflorgenin CAS:697300-41-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN76861MG
Encompass_Preferred
Medchemexpress, HY-N7686 1mg Paeoniflorgenin CAS:697300-41-7 Paeoniflorgenin is a deglucosylated metabolite of Paeoniflorin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3008