Learn More
Medchemexpress LLC Paeoniflorin sulfite | 1146967-98-7 | 10 MG

Supplier: Medchemexpress LLC HYN763910MG
Paeoniflorin sulfite (HY-N7639) is an off-white solid chemical with a purity of 99.15% (HPLC). It has a molecular formula of C₂₃H₂₈O₁₃S and a molecular weight of 544.53. This product is intended for research use only.
- Off-white solid appearance
- High purity of 99.15% (HPLC)
- Molecular formula: C₂₃H₂₈O₁₃S
- Molecular weight: 544.53
- Store at 4°C, protected from light
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.