Learn More
Medchemexpress LLC Paeoniflorin sulfite | 1146967-98-7 | 5 MG

Supplier: Medchemexpress LLC HYN76395MG
Paeoniflorin sulfite is a white to off-white solid with high purity. It is a compound that can be formed from Paeoniflorin, a main component of Paeoniae Radix Alba, during the sulfur-fumigation process of Paeoniae Radix Alba.
- Purity (HPLC): 99.15%
- Appearance: white to off-white solid
- Consistent with structure by ¹H NMR spectrum
- Molecular formula: C23H28O13S
- Molecular weight: 544.53
- Storage at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.