Learn More
Cayman Chemical PAF C18d4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10010229500 UG
Encompass_Preferred
An internal standard for the quantification of PAF C-18 by GC- or LC-MS
Catalog No. 50-288-9486