Learn More
Uline Pail Opener [ULINE]
Supplier:  Uline H1468
Encompass
A handheld steel industrial pail and drum opener lid remover with prying end and hammer head for prying off friction fit and crimped-on lids without needing other tools.
Catalog No. NC2197366