Learn More
Essendant Rust-Oleum Industrial Choice M1600 System Solvent-Based Precision Line Marking Paint, Flat White, 17 oz Aerosol Can, 12/Carton
Supplier:  Essendant RST203030V
Encompass_Preferred
Create crisp easy-to-read marks in bright ultra-durable colors with Industrial Choice M1600 System Precision Line Marking Paint Compared to common alternatives this solvent-based spray paint not only allows you to cover more linear feet per can but it also provides the best possible resistance against premature chalking and fading to ensure that marks remain vivid for much longer periods of time Aerosol can s precision tip virtually eliminates the potential for overspray and the paint can be applied in temperatures down to 25 F Use it on grass gravel packed soil concrete asphalt and more For best results apply using marking pistols or marking wands to complete everything from construction-site to athletic-field marking tasks Paint Sub-Category- Spray Paint Solvent-Based Intended Application Environment- Exterior Finish/Sheen- Flat Color(s)- White
Catalog No. 50-000-28451