Learn More
Grainger DYKEM Paint Marker, Fiber Tip, Black
Supplier:  Grainger 2GKT8
Encompass_Preferred
The DYKEM 2GKT8 Paint Marker is a fiber tip paint marker that creates durable marks on a variety of surfaces including metal, plastic, glass, and more. This paint marker features fast-drying paint that is resistant to UV rays, water, oil, and abrasion. The fiber tip applicator provides a medium line width and allows for writing at any angle. This versatile paint marker can be used for marking, labeling, and identification purposes. The paint is lead-free and AP certified.
Catalog No. NC1166376