Learn More
Essendant Rust-Oleum Commercial The MULE (Modified Urethane Latex Epoxy), Interior/Exterior, Gloss Safety Yellow, 1 gal Bucket/Pail, 2/Carton
Supplier:  Essendant RST375661
Encompass_Preferred
The MULE is an easy to use abrasion-resistant commercial coating designed specifically for high traffic areas that are subject to routine cleaning and disinfection- schools healthcare facilities offices hospitality environments and more Verstile Modified Urethane Latex Epoxy (MULE) features excellent adhesion qualities allowing it to be applied on a wide range of both primed and previously painted materials indoor and out Use it on walls doors moldings railings and even floors Low-odor water-based formula Paint Sub-Category- Epoxy Intended Application Environment- Interior/Exterior Finish/Sheen- Gloss Color(s)- Safety Yellow
Catalog No. 50-000-27408