Learn More
Essendant Zinsser Mold Killing Primer, Interior/Exterior, Flat White, 1 gal Bucket/Pail, 2/Carton
Supplier: Essendant RST276049CT
Zinsser Mold Killing Primer is a protective water-based coating that can be applied directly over existing mold mildew and odor-causing bacteria Featuring a proven registered and approved antimicrobial-fungicidal agent it not only covers over associated stains but also kills all pre-existing fungal organisms Use it directly on nearly any dry debris-free non-porous interior or exterior surface with minimal pre-cleaning in just about any residential commercial or industrial environment- homes offices buildings retail establishments schools hospitals factories and more And though this primer is compatible with any topcoat product it pairs perfectly with Zinsser PERMA-WHITE Mold and Mildew-Proof paints To fully cover over and obscure heavy stains a second primer coat with a tailored stain-blocking product may be required In such scenarios BIN and Cover Stain primers are highly recommended Paint Sub-Category- Primer Water-Based Intended Application Environment- In