Learn More
ARBOR SCIENTIFIC PAIR OF MEGA MIRRORS
SDP
Supplier:  ARBOR SCIENTIFIC P27150
Encompass
These extra-large parabolic mirrors make demonstrating optical principles easy. An example. The concave mirror can demonstrate reflection and focusing of light, heat, and sound. Discussion of shapes and uses of satellite dishes can be visually demonstrated.
Catalog No. NC0731647