Learn More
Sino Biological Peptide Substrates: PAKtide
Supplier:  Sino Biological P0858
Encompass_Preferred
The 10 amino acids of PAKtide peptide (RRRLSFAEPG) contain a serine/threonine protein kinase phosphorylation site in a common seven-residue epitope (1, 2).
Catalog No. 50-256-8388