Learn More
Cayman Chemical Pal-VGVAPGacetate 1g
SDP
Supplier:  Cayman Chemical 351851 G
Encompass_Preferred
A lipidated hexapeptide composed of an elastin fragment conjugated to palmitic acid
Catalog No. 50-291-1281