Learn More
Medchemexpress LLC HY-50767 50mg Medchemexpress, Palbociclib CAS:571190-30-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5076750MG
Encompass_Preferred
Medchemexpress, HY-50767 50mg Palbociclib CAS:571190-30-2 Palbociclib (PD 0332991) is a selective CDK4 and CDK6 inhibitor with IC50s of 11 and 16 nM, respectively. Palbociclib has the potential for ER-positive and HER2-negative breast cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9335