Learn More
Medchemexpress LLC HY-50767A 10mg Medchemexpress, Palbociclib (hydrochloride) CAS:827022-32-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50767A10MG
Encompass_Preferred
Medchemexpress, HY-50767A 10mg Palbociclib (hydrochloride) CAS:827022-32-2 Palbociclib hydrochloride is a highly selective CDK4/6 inhibitor with IC50s of 11 nM and 16 nM, respectively. Palbociclib hydrochloride has the potential for ER-positive and HER2-negative breast cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9336