Learn More
Medchemexpress LLC Palbociclib hydrochloride | 571189-11-2 | 99.96% | C24H30ClN7O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HY50767C1G
Encompass_Preferred
Palbociclib hydrochloride is an orally active, selective CDK4 and CDK6 inhibitor with IC50 values of 11 nM and 16 nM, respectively. It exhibits potent anti-proliferative activity and induces cell cycle arrest in cancer cells. This compound is suitable for research on HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
  • Selective CDK4 and CDK6 inhibitor
  • Potent anti-proliferative activity
  • Induces cell cycle arrest in cancer cells
  • Used in research of HR-positive and HER2-negative breast cancer
  • Used in research of hepatocellular carcinoma
Catalog No. 50-003-58765